Learn More
IKA™ MATRIX Attachment
Attachment design ensures an optimal temperature transfer to guarantee valid results.
Brand: IKA™ 0030000821
This item is not returnable.
View return policy
Description
- Thermoshakers automatically recognize which attachment is connected to the device via an interface in the locking mechanism
- Attachments can be changed quickly and easily with an ergonomic lever
- As part of IKA's Integrated Safety Concept, the insulating plastic coating of the attachment ensures user safety, as high temperatures are not transferred to the frame
- Suitable for use with MATRIX Orbital Delta Plus, and MATRIX Delta Plus
Specifications
Conical Vials | |
Attachment for Conical Vials | |
Blood Samples, Pharmaceutical Agents,/DNA/RNA Samples | |
8 |
15 mL | |
150 x 141 x 53 mm | |
MATRIX Orbital Delta Plus, MATRIX Delta Plus |