Learn More
MBL™ Borosilicate Glass Class A Volumetric Flask with Stopper
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
Height (Metric):
70 mm
90 mm
110 mm
140 mm
170 mm
210 mm
220 mm
260 mm
300 mm
370 mm
475 mm
Unit Size:
Each
Pack of 2
Pack of 5
Description
- Finely calibrated to Class A tolerances
- Each flask is laser etched with an individual serial number, batch code and date of manufacture
- Conformity 'H' certified
- Inscriptions in blue enamel
- Supplied with polyethylene stopper
- Batch certificate available to download online
Spécification
Spécification
| Certifications/Compliance | Class A, ISO 1042, DIN 12664, Conformity 'H' certified |
| Closure Material | Polyethylene |
| Description | Blue enamel inscriptions |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 300 mm |
| Tolerance | ±0.4 mL |
| Graduated | Yes |
| Class | A |
| Closure Size | 24/29 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu