missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCTS1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99158H
This item is not returnable.
View return policy
Description
MCTS1 Polyclonal specifically detects MCTS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MCTS1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
malignant T cell amplified sequence 1, Malignant T-Cell-Amplified Sequence 1, MCT-1, Multiple Copies T-Cell Malignancies | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human MCTS1. | |
0.1 mL | |
Cardiovascular Biology | |
28985 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction