Learn More
Kartell™ Measuring Scoops
Click to view available options
Capacity (Metric):
10 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Pack of 6
Description
Shaped conveniently for sampling
Specifications
Specifications
| Capacity (English) | 3.38 oz. |
| Capacity (Metric) | 100 mL |
| Product Type | Measuring Scoop |
| Dimensions (L x W) | 67.2 x 201.7 x 34 mm |
| Material | Polypropylene |