Learn More
Binder™ Medium Shelf Set for Environmental Walk-in Chambers
Shop All BINDER Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Accessory for WIC models 2 and 2 C
Specifications
Specifications
| Color | Silver |
| For Use With (Equipment) | Binder™ Environmental Walk-in Chambers 2 and 2C |
| Material | Stainless Steel |
| Type | Shelf Set |