missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ VITLAB™ Melamine Formaldehyde Instrument Tray

Support your laboratory instruments, tools, and sensitive utensils using the VITLAB™ Melamine Formaldehyde Instrument Tray, which is stable and self-supporting.

Brand:  BrandTech™ 71898

 View more versions of this product

Product Code. 10070581

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • White Melamine
  • Flat shape
  • Rounded corners
  • Smooth surface
  • Easy to clean
Specifications

Specifications

355 x 240 mm
17 mm
Melamine Formaldehyde
240 mm
Flat
355 x 240 x 17 mm
355 mm
Instrument Tray
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers