missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ VITLAB™ Melamine Formaldehyde Instrument Tray
Support your laboratory instruments, tools, and sensitive utensils using the VITLAB™ Melamine Formaldehyde Instrument Tray, which is stable and self-supporting.
Brand: BrandTech™ 71998
This item is not returnable.
View return policy
Description
- White Melamine
- Flat shape
- Rounded corners
- Smooth surface
- Easy to clean
Specifications
428 x 288 mm | |
White | |
428 mm | |
Instrument Tray | |
1 Each |
428 x 288 x 17 mm | |
17 mm | |
Polyurethane | |
288 mm | |
Rectangular |