missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ VITLAB™ Melamine Formaldehyde Instrument Tray
Support your laboratory instruments, tools, and sensitive utensils using the VITLAB™ Melamine Formaldehyde Instrument Tray, which is stable and self-supporting.
Brand: BrandTech™ 72498
This item is not returnable.
View return policy
Description
- White Melamine
- Flat shape
- Rounded corners
- Smooth surface
- Easy to clean
Specifications
340 x 245 x 100 mm | |
100 mm | |
Polyurethane | |
245 mm | |
Rectangular |
White | |
340 mm | |
Instrument Tray | |
1 Piece |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction