missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene-2-sulfonyl hydrazide, 97%, Thermo Scientific™

Product Code. 15411898
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15411898

Brand: Thermo Scientific Alfa Aesar H55586.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16182-15-3
Molecular Formula C9H14N2O2S
Molecular Weight (g/mol) 214.283
MDL Number MFCD00007585
InChI Key JQUBKTQDNVZHIY-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
PubChem CID 85321
IUPAC Name 2,4,6-trimethylbenzenesulfonohydrazide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C

Specifications

Melting Point 112°C to 114°C (decomposition)
Color White
Quantity 25 g
UN Number UN1325
Beilstein 2118380
Formula Weight 214.28
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Mesitylene-2-sulfonyl hydrazide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.