missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldicarb in Acetonitrile 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule
Specifications
Specifications
Concentration | 10μg/mL |
CAS | 116-06-3 |
Packaging | Ampoule |
Quantity | 10 mL |
Type | Organic Single-component Standard Solution |
Molecular Formula | C7H14N2O2S |
UN Number | 1648 |
InChI Key | QGLZXHRNAYXIBU-WEVVVXLNSA-N |
SMILES | CC(C)(C=NOC(=O)NC)SC |
IUPAC Name | [(E)-(2-methyl-2-methylsulfanylpropylidene)amino] N-methylcarbamate |
Show More |
Safety and Handling
Product Identifier
- Standard solution Aldicarb,2-methyl-2-(methylthio)propanal-O-(N-methylcarbamoyl)oxime 10ug/ml in acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H311-Toxic in contact with skin.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'