Learn More
SI Analytics™ ScienceLine Metal Combination Electrode
SI Analytics™ ScienceLine Metal Combination Electrode is the first choice for titration and can be used in a range of applications like general redox titrations.
Brand:  SI Analytics™ 285102413
291.13 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 10478952
  • 357.00 EUR / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
  • Metal combination electrode with plug head and connection cable
  • For use with determining acid value and halogenide (mostly chloride)
  • For determinging iodine number, peroxice value, and general redox titrations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plug Head
Glass
Halogenide Titrations
Agcl 62
120 mm
4 mm
Platinum Diaphragm
Metal Combination Electrode
Ag/AgCl
-5°C to +100°C
12 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products