Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Custom Needles with Point 2, 3 or 4
Click to view available options
Needle Gauge:
20 G
Unit Size:
Pack of 6
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Nickel-plated Brass (Hub), Stainless Steel |
| Needle Gauge | 20 G |
| Diameter (Metric) Inner | 0.60 mm |
| Diameter (Metric) Outer | 0.91 mm |
| Product Type | Luer Lock Custom Needle |
| Quantity | 6/Pk. |
| Length (Metric) Needle | 9.52 to 304 mm |
| Needle Point Style | 2, 3, or 4 |
| For Use With (Equipment) | TLL Syringes and LT or TLL Connectors |
| Show More |