missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metanil Yellow

28.84 EUR - 83.11 EUR

Chemical Identifiers

CAS 587-98-4
Molecular Formula C18H14N3NaO3S
Molecular Weight (g/mol) 375.378
MDL Number MFCD00007486
InChI Key NYGZLYXAPMMJTE-UHFFFAOYSA-M
Synonym C.I. 13065
PubChem CID 3935589
ChEBI CHEBI:87235
IUPAC Name sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478147
View Documents
Thermo Scientific Chemicals
A17527.18
50 g
28.84 EUR
50g
In Stock
to see stock.
Add to basket
 
11488147
View Documents
Thermo Scientific Chemicals
A17527.30
250 g
83.11 EUR
250g
In Stock
to see stock.
Add to basket
 
Description

Description

Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

Solubility
Soluble in water (25g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

587-98-4
375.378
NYGZLYXAPMMJTE-UHFFFAOYSA-M
3935589
sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
C18H14N3NaO3S
MFCD00007486
C.I. 13065
CHEBI:87235
C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]

Specifications

587-98-4
MFCD00007486
14,5928
Soluble in water (25g/L).
C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+]
375.378
CHEBI:87235
Odorless
Metanil Yellow
C18H14N3NaO3S
3831568
C.I. 13065
NYGZLYXAPMMJTE-UHFFFAOYSA-M
sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate
3935589
375.38
10
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H361

missing translation for 'einecsNumber' : 209-608-2

missing translation for 'rtecsNumber' : DB7329500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.