missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Dried, Karl Fischer Reagent, Scharlab

Product Code. 11328444
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in solvents, the synthesis of organic products, in antifreeze compositions, and in solvents for animal and vegetable oils extractions.

  • Synonyms: Methyl alcohol, Carbinol, Methynol, Wood alcohol
  • Molecular Formula: CH3OH
  • Molar Mass: 32.04g/mol
  • CAS: 67-56-1
  • EIN ECS No.: 200-659-6
  • Dielectric Constant: 32.6 (at 25°C)

Specifications

Melting Point -98°C
CAS 67-56-1
Acidity 0.0003mEq/g max
Alkalinity 0.0002mEq/g max
Appearance Clear
Density 0.79g/cm3 
Boiling Point 64.5°C
Quantity 1 L
Flash Point 11°C
Identification Passes Test (IR spectrum)
Ignition Point 455°C
Physical Form Dried
Refractive Index 1.3288 (n 20°C/D)
Residue after Evaporation 0.0005% max
Molecular Formula CH4O
MDL Number MFCD00004595
Vapor Pressure 128hPa (at 20°C)
Water 0.005% max
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Miscible in water at 20°C
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity 99.8% min (G.C.)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol, dried (max. 0,005% H2O), reagent grade (Karl Fischer)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.