Learn More
Phenolics Mix B, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 2,3,4,6-Tetrachlorophenol | 58-90-2 | 2000μg/mL |
| 2,4,5-Trichlorophenol | 95-95-4 | 2000μg/mL |
| 2,6-Dichlorophenol | 87-65-0 | 2000μg/mL |
| 2-Methylphenol | 95-48-7 | 2000μg/mL |
| 3-Methylphenol | 108-39-4 | 2000μg/mL |
| 4-Methylphenol | 106-44-5 | 2000μg/mL |
| Dinoseb | 88-85-7 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Phenolics Mix B |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA Methods 8040, 8041 |
| Analyte or Component Names | 2,6-Dichlorophenol; Dinoseb; 2-Methylphenol; 3-Methylphenol; 4-Methylphenol; 2,3 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Element | 2,6-Dichlorophenol, Dinoseb, 2-Methylphenol, 3-Methylphenol, 4-Methylphenol |
| Solvent or Matrix | Isopropanol |
| Quantity | 1 mL |
Safety and Handling
Product Identifier
- Phenolics Mix B
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : propan-2-ol, 2,4,5-trichlorophenol, 2,6-dichlorophenol, dinoseb, 2,3,4,6-tetrachlorophenol
Product intended for laboratory use only