missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic acid, 97+%, Thermo Scientific Chemicals

26.73 EUR - 532.65 EUR

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004308
InChI Key RMIODHQZRUFFFF-UHFFFAOYSA-N
Synonym methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
PubChem CID 12251
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10781461
View Documents
Thermo Scientific Acros
186770250
25 mL Glass bottle
26.73 EUR
25mL
Please to purchase this item. Need a web account? Register with us today!
10781651
View Documents
Thermo Scientific Acros
186772500
250 mL Glass bottle
129.65 EUR
250mL
Please to purchase this item. Need a web account? Register with us today!
10072310
View Documents
Thermo Scientific Acros
186770010
1 L Glass bottle
532.65 EUR
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

625-45-6
90.08
RMIODHQZRUFFFF-UHFFFAOYSA-N
12251
COCC(=O)O
C3H6O3
MFCD00004308
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
2-methoxyacetic acid

Specifications

625-45-6
Colorless to Yellow
202°C to 204°C
Authentic
C3H6O3
1.4153 to 1.4173
03, 232
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
RMIODHQZRUFFFF-UHFFFAOYSA-N
2-methoxyacetic acid
12251
97+%
Methoxyacetic acid, 98%
7°C
1.1740g/mL
121°C
97+%
CH3OCH2CO2H
MFCD00004308
1.174
Solubility in water: freely soluble
COCC(=O)O
90.08
90.08
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-894-6

missing translation for 'rtecsNumber' : AI8650000

missing translation for 'tsca' : TSCA

Product Identifier
  • Methoxyacetic acid

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only