missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 98%, Thermo Scientific™

Product Code. p-4813549 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15224854

Brand: Thermo Scientific Acros 450770050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 85006-31-1
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00051822
InChI Key YICRPERKKBDRSP-UHFFFAOYSA-N
Synonym methyl3-amino-4-methylthiophene-2-carboxylate, 3-amino-4-methylthiophene-2-carboxylic acid methyl ester, unii-3la24473as, 3-amino-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, methyl 3-amino-4-methyl-2-thenoate, methyl 3-amino-4-methyl-2-thiophenecarboxylate, methyl-3-amino-4-methylthiophene-2-carboxylate, methyl 3-amino-4-methyl-thiophene-2-carboxylate, 3-amino-4-methyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 123584
IUPAC Name methyl 3-amino-4-methylthiophene-2-carboxylate
SMILES COC(=O)C1=C(N)C(C)=CS1

Specifications

Melting Point 85°C to 88°C
Flash Point >100°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 171.22
Percent Purity 98%
Chemical Name or Material Methyl 3-amino-4-methyl-2-thiophenecarboxylate
compliance-icons
Product Identifier
  • Methyl 3-amino-4-methylthiophene-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.