Learn More
Emma Safety Footwear Metric Safety Shoes
Product Code. 15948551
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15948551
Brand: Emma Safety Footwear MM50654047
Only null left
Add to basket
Add to basket
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Elastic
Color White
Design Metric
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 47
Toe Material Steel