Learn More
Metro™ Flexline™ Cart Accessories
Click to view available options
Dimensions (L x W x H):
8.5 x 4.5 x 16.75 in.
Color:
Gray
Capacity (English):
7 gal.
Product Type:
Waste Basket and Holder
Unit Size:
Each
Description
- 28 quart
- Dimensions: 37 x 27.5 x 39cm (14.5 x 10.875 x 15.375 in.)
- Holder included
Specifications
Specifications
| Capacity (English) | 7 gal. |
| Product Type | Waste Basket and Holder |
| Capacity (Metric) | 26.5 L |
| Color | Gray |
| Dimensions (L x W x H) | 8.5 x 4.5 x 16.75 in. |
| For Use With (Equipment) | Flexline and Lifeline Carts |
| Height (English) | 16.75 in. |
| Height (Metric) | 42.54 cm |
| Includes | Holder |
| Length (English) | 8.5 in. |
| Show More |