Learn More
Metro™ MetroMax iQ™ Stationary Security Storage Unit
Protect sensitive materials from loss or pilferage. Stationary security storage unit allows for protection in a secure, parked unit within your work environment.
Brand:  Metro™ MQSEC55E
 View more versions of this product
Product Code. 15997161
  • 2595.00 EUR / Each
Estimated Shipment: 24-02-2025
Description
  • Heavy gauge open wire construction keep contents visible for easy inventory check
  • Built-in Microban™ antimicrobial product protection to keep the product cleaner between cleanings
  • Double door with ergonomic -+-turn handle
  • Each door opens 270° and can be secured to the side panels
  • Posts have leveling feet to compensate for uneven surfaces
Specifications
Specifications
Stationary Security Unit
50.875 x 26.9375 x 66.1875 in.
Epoxy-coated Steel
Taupe
24 x 48 in. Shelf
2