Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Product Code. 15926621
Click to view available options
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
Unit Size:
Each
Product Code. 15926621
Brand: Metro™ 2130NFS
Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.
  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Product Type Solid Shelf
Color Silver
Material Stainless Steel
Autoclavable Autoclavable
Dimensions (L x W) 30 x 21 in. (76 x 53 cm)
For Use With (Equipment) Metro Super Erecta Shelving System
Length (English) 30 in.
Length (Metric) 76 cm
Width (English) 21 in.
Width (Metric) 53 cm