Learn More
Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable
Click to view available options
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
60 in.
Unit Size:
Each
Description
- All-stainless construction
- Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications
Specifications
| Product Type | Solid Shelf |
| Color | Silver |
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| Dimensions (L x W) | 30 x 18 in. (76 x 45.7 cm) |
| For Use With (Equipment) | Metro Super Erecta Shelving System |
| Length (English) | 30 in. |
| Length (Metric) | 76 cm |
| Width (English) | 18 in. |
| Width (Metric) | 45.7 cm |