Learn More
Mettler Toledo™ Infrared Adapter For Densito™ 30PX
Accessory for use with Mettler Toledo 30PX Density/Gravity Meters
Brand: Mettler Toledo™ 51325006
642.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For data transmission
- 1/Pk.
Specifications
Infrared-Adapter (RS 9pin, male) | |
Infrared Adapter | |
Data Transmission |
English (US)French | |
Densito 30PX Portable Specific Gravity/Density Meter |
Product Content Correction
Product Title
Mettler Toledo™ Infrared Adapter For Densito™ 30PX > Densito 30PX Infrared Adapter
Spot an opportunity for improvement?Share a Content Correction