missing translation for 'onlineSavingsMsg'
Learn More

MF498, MedChemExpress

Product Code. 16404168
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16404168

Brand: Medchem Express HY10794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MF498 is a novel and selective E prostanoid receptor 4 (EP4 receptor) antagonist, displayed strong binding affinity for the EP4 receptor with Ki of 0.7 nM.

MF498 is a novel and selective E prostanoid receptor 4 (EP4 receptor) antagonist, displayed strong binding affinity for the EP4 receptor with Ki of 0.7 nM.

Specifications

Chemical Name or Material MF498
CAS 915191-42-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H33N3O7S
Quantity 10 mg
Solubility Information DMSO : ≥ 31 mg/mL (51.35 mM)
SMILES O=C1N(C2=C(C)C=C(CS(NC(CC3=CC=CC=C3OC)=O)(=O)=O)C=C2)CC4=C(OCC)C5=NC=CC=C5C(OCC)=C41
Molecular Weight (g/mol) 603.69
Formula Weight 603.69
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.