Learn More
Eppendorf™ Centrifuge 5430/5430 R
GREENER_CHOICE
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
Brand:  Eppendorf™ 5428000010
6160.13 EUR valid until 2024-12-06
Use promo code "21633" to get your promotional price.
 View more versions of this product
Product Code. 15825073
  • 8055.00 EUR / 1 set
Description
  • Max. rotor capacity: 48 × 1.5/2.0 mL, 6 × 50 mL, 2 × MTP
  • Max. speed: 30,130 × g (17,500 rpm)
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock™ – System for quickly opening and closing the rotor lid
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    30 x 2 mL
    30,130 x g
    Large, brightly lit LCD
    29 cm
    Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
    15 in.
    1.050 W
    With Rotor FA-45-30-11 incl. rotor lid
    230 V, 50/60 Hz (EU)
    EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 2 EN 50581 (refer to manual for more details)
    50/60 Hz
    Centrifuge 5430 R, keypad
    17,500 rpm
    Yes
    11.7 in.
    <54 dB(A) with Rotor F-45-30-11
    30 sec. to 99:59 hrs., with continuous run function
    38 cm
    UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    380 x 640 x 290 mm
    Yes
    -11°C to +40°C
    230 V