Learn More
Ansell™ Microchem™ CFR Coveralls with Hood
MICROCHEM™ CFR is a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX™, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
1320.00 EUR - 1650.00 EUR
Specifications
Product Type | Protective Coveralls |
---|---|
Material | Flame-retardant Fabric with PVC Barrier Film |
Color | Red |
Ankle Style | Elastic |
Cuff Style | Elastic |
Alert:
Microchem™ CFR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15546484
|
Ansell™
RD96T-00111-03 |
Medium |
1650.00 EUR
Pack of 25 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
15556484
|
Ansell™
RD96T-00111-04 |
Large |
1650.00 EUR
Pack of 25 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15566484
|
Ansell™
RD96T-00111-05 |
X-Large |
1650.00 EUR
Pack of 25 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15586484
|
Ansell™
RD96T-00111-07 |
3X-Large |
1320.00 EUR
Pack of 20 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
15576484
|
Ansell™
RD96T-00111-06 |
2X-Large |
1320.00 EUR
Pack of 20 |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
Description
- Coverall with 2 piece hood
- Elasticated hood, wrists and ankles
- Double zip closure
- Flame retardant fabric treated with PVC barrier film
- Protects from liquid chemicals to EN Type 3 and 4
- Optimized body fit provides comfort and safety
- Highly visible bright red colour to improve worker safety
Specifications
Protective Coveralls | |
Red | |
Elastic | |
2-panel, Elastic | |
Stitched, Taped | |
None |
Flame-retardant Fabric with PVC Barrier Film | |
Elastic | |
Hooded | |
Double Zipper | |
EN14605 Type 3 and 4, EN ISO 13982-1 Type 5, EN 1073-2, EN ISO 14116, EN 1149-5 | |
Includes Hood |