Learn More
Invitrogen™ Microplates for Fluorescence-based Assays
Product Code. p-200089241
Click to view available options
No. of Wells:
96
Quantity:
10 Plates
30 Plates
Unit Size:
Pack of 10
Pack of 30
Product Code. 15329413
Brand: Invitrogen™ M33089
Only null left
Add to basket
Add to basket
Microplates for fluorescence-based assays are 96- and 384-well, black-walled, clear-bottom microplates that are optimized for fluorescence-based applications on standard fluorescence microplate readers.
Microplates for fluorescence-based assays are black-walled, clear-bottom microplates that minimize well-to-well crosstalk and autofluorescence. These 96- and 384-well microplates are compatible with standard fluorescence microplate readers and optimized for fluorescence-based applications including nucleic acid quantitation (e.g., Quant-iT assays, including PicoGreen, RiboGreen, and OliGreen).
Features of the microplates for fluorescence-based assays include:
  • Black-walled design—minimizes well-to-well crosstalk and autofluorescence
  • Clear-bottom wells—allows for optimal fluorescence detection
  • Compatibility—suitable for standard fluorescence microplate readers
  • Optimized for fluorescence microplate assays—ideal for nucleic acid quantitation (e.g., Quant-iT assays)
  • Versatile formats—available in 96- and 384-well configurations
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Content And Storage Store at room temperature.
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 10-plates
Product Line Invitrogen
Color Clear Bottom, Black
Disposable Single-use
Product Type Microplates
For Use With (Equipment) Microplate Reader
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 200 μL
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 10 Plates