Learn More
Ohaus™ Microtube Holder Kit
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Microtubes
Unit Size:
Each
Description
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
| Product Type | Microtube and Well Plate Holder Kit |
| Material | Foam |
| For Use With (Equipment) | Microtubes |