Learn More
Ohaus™ Microtube Holder Kit
Product Code. 15702619 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Microtubes
Unit Size:
Each
Product Code. 15702619
Brand: Ohaus™ 30400198
Only null left
Add to basket
Add to basket
OHAUS™ Microtube Holder Kit for effortlessly mixing microtubes and microplates.
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microtube and Well Plate Holder Kit
Material Foam
For Use With (Equipment) Microtubes