missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Mini Tube System

Product Code. 11376324 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
0.65 mL
1.1 mL
12 Cap Strip
8 Cap Strip
Sterility:
Non-sterile
Sterile
Packaging:
12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
120 Strips/Bag; 5 Bags/Cs.
120 Strips/PK., 5 Packs/Cs.
8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs.
80 Strips/Bag; 5 Bags/Cs.
80 Strips/PK., 5 Packs/Cs.
96/Rack, 10 Racks/Pk., 5 Packs/Cs.
960 Tubes/Bag, 5 Bags/Cs.
Unit Size:
Pack of 120
Pack of 400
Pack of 4800
Pack of 600
Pack of 80
Pack of 960
This item is not returnable. View return policy

Product Code. 11376324

Brand: Axygen™ MTS8CPCS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polypropylene tube system for compound library storage

Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.

  • Rack features a one-way lid design and alpha-numeric grid
  • Molded-in condensation rings on the lid minimize cross contamination
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 8 Cap Strip
Color Clear
Material Polyethylene
Format Strip
For Use With (Application) Ensures positive sample identification
No. per Pack 120/Pk.
Packaging 120 Strips/PK., 5 Packs/Cs.
Sterility Sterile
Bottom Shape Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.