Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Nylon and Silicone
Versatile check valves can be installed in any position
6.05 EUR
Product Code | Brand | Inlet | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Inlet | Price | Quantity & Availability | |||||
30053887
|
Cole-Parmer
50115-68 |
3/16 in. hose barb |
6.05 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30054057
|
Cole-Parmer
50115-19 |
1/4 in. hose barb |
6.05 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30052932
|
Cole-Parmer
50115-18 |
1/8 in. hose barb |
6.05 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressureSpot an opportunity for improvement?Share a Content Correction