Learn More
Bryson Packaging™ Polyethylene Bags
Product Code. 10031474
Click to view available options
Width (Metric):
40 mm
60 mm
75 mm
90 mm
100 mm
115 mm
125 mm
150 mm
190 mm
230 mm
255 mm
330 mm
Height (Metric):
65 mm
75 mm
80 mm
115 mm
140 mm
175 mm
190 mm
230 mm
325 mm
355 mm
457 mm
Color:
Clear
Clear with White Bars
Unit Size:
Pack of 100
Product Code. 10031474
Brand: Bryson Packaging™ BAJ360120S
Only null left
Add to basket
Add to basket
Transparent and excellent for shipping, storage and as sample bags.
  • Available in a wide range of sizes/capacities
  • With resealable strips
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Width (Metric) 125 mm
Height (Metric) 190 mm