Learn More
Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags
Product Code. 11358203
Click to view available options
Width (Metric):
57 mm
60 mm
90 mm
115 mm
125 mm
150 mm
230 mm
254 mm
279 mm
Height (Metric):
60 mm
76 mm
115 mm
190 mm
230 mm
325 mm
356 mm
406 mm
Unit Size:
Pack of 100
Product Code. 11358203
Brand: Bryson Packaging™ BAJ340140D
Only null left
Add to basket
Add to basket
Store and send samples and stationary in resealable bags. Bryson Packaging™ Minigrip™ Polyethylene Write-on Bags are clear polyethylene bags with three white bars for labelling. Square and rectangular bags are available in a range of sizes. 100 and 1,000 packs.
  • Clear polyethylene bags with Grip Seal resealable strips
  • Three generously sized write-on labelling bars for easy identification and organization
  • Ideal for storing stationery and sending samples
  • Available in a range of sizes to meet storage needs
Specifications
Description Resealable Write-On Bag
Color Clear with White Bars
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Thickness (Metric) 50 μm
Quantity 100 Pack
Width (Metric) 57 mm
Height (Metric) 76 mm