Learn More
Cytiva Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system with minimal user intervention.
Brand: Cytiva OA010C12
421.80 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Efficient processing
- This product is not approved for use in medical, clinical, surgical or other patient applications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
PES | |
Non-sterile | |
1/Pk. |
10 kDa | |
50 cm2 |