missing translation for 'onlineSavingsMsg'
Learn More

Minocycline Hydrochloride, MP Biomedicals™

Product Code. 11434911
Click to view available options
Quantity:
100 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
Synonym minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino
PubChem CID 54685925
IUPAC Name hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 493.9
Physical Form Powder
Chemical Name or Material Minocycline Hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'