Learn More
Mirabegron, MedChemExpress
Product Code. 16496158
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16496158
Brand: Medchem Express HY14773
MedChemExpress Mirabegron is a selective β3-adrenoceptor agonist with EC50 of 22.4 nM.
Mirabegron is a selective β3-adrenoceptor agonist with EC50 of 22.4 nM.
Specifications
Chemical Name or Material Mirabegron
Physical Form Powder
CAS 223673-61-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H24N4O2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym YM178
Solubility Information DMSO : 100 mg/mL (252.20 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(CCNC[C@H](O)C2=CC=CC=C2)C=C1)CC3=CSC(N)=N3
Molecular Weight (g/mol) 396.51
Formula Weight 396.51
Percent Purity 98.98%
Grade Research