Learn More
Fisherbrand™ Mixed Heating (Steam or Electrical)
Mixed heating from a steam source for water heating and an electrically powered source for drying.
Specifications
For Use With (Application) Steam or Electrical Heating
View More Specs
Description
Description
  • The energy for water heating comes from steam source
  • The energy for drying is electrically powered
  • If the steam fails, the machine continues to run on electrical power
  • The machine can be programmed to run on steam in certain periods and electricity at others
Specifications
Specifications
Steam or Electrical Heating