missing translation for 'onlineSavingsMsg'
Learn More

ML 141, Tocris Bioscience™

Product Code. 18035812
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective Cdc42 Rho family inhibitor

Allosteric inhibitor of Cdc42 GTPase (EC50 = 2.1 μM). Selective for Cdc42 over other members of the Rho GTPase family including Rac1, Rab2 and Rab7. Inhibits ovarian cancer cell migration in vitro.

Chemical Identifiers

CAS 71203-35-5
Molecular Formula C22H21N3O3S
Molecular Weight (g/mol) 407.488
InChI Key QBNZBMVRFYREHK-UHFFFAOYSA-N
Synonym 4-3-4-methoxyphenyl-5-phenyl-3,4-dihydropyrazol-2-yl benzenesulfonamide, 4-5-4-methoxyphenyl-3-phenyl-4,5-dihydro-1h-pyrazol-1-yl benzenesulfonamide, 4-5-4-methoxyphenyl-3-phenyl-4,5-dihydropyrazol-1-yl benzenesulfonamide, cid, ml hplc, 4-5-4-methoxyphenyl-3-phenyl-2-pyrazolinyl benzenesulfonamide, 4-5-4-methoxyphenyl-3-phenyl-2-pyrazolin-1-yl benzenesulfonamide, 4-5-4-methoxyphenyl-3-phenyl-4,5-dihydro-1hpyrazol-1-yl benzenesulfonamide, 4-4,5-dihydro-5-4-methoxyphenyl-3-phenyl-1h-pyrazol-1-yl benzenesulfonamide, 4-5-4-methoxyphenyl-3-phenyl-4,5-dihydro-1h-pyrazol-1-yl benzene-1-sulfonamide
PubChem CID 2950007
IUPAC Name 4-[3-(4-methoxyphenyl)-5-phenyl-3,4-dihydropyrazol-2-yl]benzenesulfonamide
SMILES COC1=CC=C(C=C1)C2CC(=NN2C3=CC=C(C=C3)S(=O)(=O)N)C4=CC=CC=C4

Specifications

Quantity 10 mg
Formula Weight 407.49
Percent Purity >99%
Chemical Name or Material Rho GTPase Inhibitors
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'