missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ML171, MedChemExpress
MedChemExpress ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay.
66.15 EUR - 132.30 EUR
Specifications
Chemical Name or Material | ML171 |
---|---|
CAS | 6631-94-3 |
Color | Orange |
Purity Grade Notes | Research |
Molecular Formula | C14H11NOS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16481646
|
Medchem Express
HY-12805 |
500 mg |
132.30 EUR
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16465986
|
Medchem Express
HY-12805 |
100 mg |
66.15 EUR
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ML171 (2-AcetylphenothiazineSpecifications
ML171 | |
Orange | |
C14H11NOS | |
DMSO : ≥ 64 mg/mL (265.22 mM) | |
241.31 | |
99.9% | |
Solid |
6631-94-3 | |
Research | |
2-Acetylphenothiazine 2-APT | |
CC(C1=CC=C2SC3=CC=CC=C3NC2=C1)=O | |
241.31 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only