missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLKL Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
191.14 EUR - 425.47 EUR
Specifications
Antigen | MLKL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18302405
|
Bio-Techne
NBP3-16934-20UL |
20 μg |
191.14 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331664
|
Bio-Techne
NBP3-16934-100UL |
100 μg |
425.47 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLKL Polyclonal antibody specifically detects MLKL in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MLKL | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
A synthetic phosphorylated peptide around S454 of human MLKL (NP_689862.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Necroptosis, Protein Kinase | |
PBS, 50% glycerol, pH7.3 | |
197259 | |
IgG | |
Affinity purified |