Learn More
Tocris Bioscience™ MM 102
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
MM 102 is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). Inhibits MLL1 methyltransferase activity and MLL-1-induced HoxA9 and Meis-1 gene expression in leukemia cells expressing the MLL1-AF9 fusion gene. Also inhibits cell growth and induces apoptosis in leukemia cells harbouring MLL1 fusion proteins.
Specifications
Specifications
| CAS | 1883545-52-5 |
| Quantity | 10 mg |
| Product Type | MM 102 |
| Purity | >98% |
| Molecular Weight (g/mol) | 783.83 |
For Research Use Only