Learn More
Köttermann Mobile Cabinet with One Lockable Door
Product Code. 17322109
Click to view available options
Hinge Side:
Right
No. of Drawers:
1
None
No. of Shelves:
1
None
Dimensions (D x W x H):
516 x 450 x 590 mm
516 x 450 x 740 mm
516 x 600 x 590 mm
516 x 600 x 740 mm
Unit Size:
Each
Product Code. 17322109
Brand: Köttermann 31500072
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Mobile Cabinet with One Lockable Door offers versatile storage space with a single door to protect stored items.
  • Versatile storage space
  • Mobile with lockable wheels
  • Features one door with locking mechanism to protect stored items
  • Many sizes and configurations available
Specifications
Color White
Cabinet Type Mobile
Dimensions (D x W x H) 516 x 600 x 740 mm
Door Type Hinged
Finish Epoxy Coated
Frame Material Steel
Hinge Side Right
Locking Mechanism Yes
No. of Doors 1
No. of Drawers None
No. of Shelves 1
Warranty None
Wheel Type Plastic with Brakes
With Casters Yes
Material Steel