Learn More
Invitrogen™ D-Myo-Inositol 1,4,5-Triphosphate, P4(5)-(1-(2-Nitrophenyl)ethyl) Ester, tris(triethylammonium) Salt (NPE-Caged Ins 1,4,5-P3)
Flash photolysis of the caging group yields highly localized release of the free nucleotide
Brand:  Invitrogen™ I23580
Product Code. 11544896
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
NPE-caged inositol 1,4,5-triphosphate is biologically inactive before photolysis. Flash photolysis of the caging group with UV illumination at 355nm results in a rapid and highly localized release of free INS 1,4,5-P3 at the site of illumination.
Cell Analysis, Cell Structure, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.