Learn More
Invitrogen™ DPX (p-Xylene-Bis-Pyridinium Bromide)
Cationic quencher
Brand: Invitrogen™ X1525
331.55 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
DPX (p-xylene-bis-pyridinium bromide) can be used in membrane fusion or permeability assays with the polyanionic, polar tracer dye ANTS (A350).
Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
1 g | |
C18H18Br2N2 | |
DPX Quencher |
Room Temperature | |
Solid |