missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fura-4F, Pentapotassium Salt, cell impermeant
Intracellular calcium indicator similar to fura-2 but with response range shifted for detection of higher Ca2+ concentrations
Brand: Invitrogen™ F14174
This item is not returnable.
View return policy
Description
- water-soluble salt form
- Useful for intracellular loading by microinjection, infusion from patch pipet or uptake induced by Influx™ pinocytic cell-loading reagent Cat. No. I-14402)
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Fluorescence Microscope | |
Fluorescent Dye-Based |
Fluorescence | |
Calcium Indicator | |
Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction