Learn More
Carcino Embryonic Antigen Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native Carcino Embryonic Antigen.
Brand:  Abnova MAB1395.1mg
Product Code. 16029809
  • 391.00 EUR / 1mg
Specifications
Carcino Embryonic Antigen
Monoclonal
Mouse monoclonal antibody raised against native Carcino Embryonic Antigen.
In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Native purified human Carcino Embryonic Antigen.
RUO
This antibody is specific to human CEA. No cross-reactivity with human NCA, NCA2 or BGP1 (Biliary glycoprotein-l).
Liquid
Immunohistochemistry
Unconjugated
The optimal working dilution should be determined by the end user.
Mouse
1 mg
Primary
Store at 4°C.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions