missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Mouse IgG1[a], Clone: 10.9, biotin, Secondary Antibody, BD
Brand: BD Biosciences 553500
469.39 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
This antibody is specific for mouse IgG1 of the Igh-C[a] and the c, d, e, f, g, h, n, o, and p haplotypes (e.g. BALB/c, C58, A, AKR, CBA, C3H, DBA/1, DBA/2, NZB, SWR). It has been reported not to react with IgG1 of the Igh-C[b] haplotype (e.g. C57BL/6, C5
Host Species: Mouse
Clone: 10.9
Isotype: IgG2a, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry
Specifications
Mouse anti-Mouse IgG1[a] | |
10.9 | |
Biotin | |
0.5 mg | |
Immunology | |
Mouse |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction