missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAGE Mouse anti-Human, Clone: 176902, R&D Systems™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: R&D Systems MAB11451-SP

Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AGER Monoclonal specifically detects AGER in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Spécification
RAGE | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q15109 | |
AGER | |
Mouse myeloma cell line NS0-derived recombinant human RAGE Gln24-Ala344 Accession # Q15109 | |
25 μg | |
Primary | |
Detects human RAGE in ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse RAGE and recombinant rat RAGE is observed. | |
Human | |
Purified |
Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
176902 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5891 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu