missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-3 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 250-41-1MG
This item is not returnable.
View return policy
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
Functional Assay | |
4.6 | |
1 mg | |
-20°C | |
<1 EU/μg | |
Mouse | |
Recombinant | |
Lyophilized |
protein with no preservative | |
Mouse BD-3 | |
E. coli | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Unconjugated | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction