missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL22/MDC Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB439
This item is not returnable.
View return policy
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot.
Specifications
| CCL22/MDC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6367 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 158113 | |
| Western Blot 1 ug/mL | |
| O88430 | |
| CCL22 | |
| E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # O88430 | |
| 500 μg | |
| Primary | |
| Detects mouse CCL22/MDC in direct ELISAs and Western blots. Shows 12% cross-reactivity with recombinant human (rh) CCL22 and recombinant mouse (rm) MIP-3 alpha , 25% cross-reactivity with rmCTACK, 6% cross-reactivity with rhBLC/BCA-1, and no cross-reactivity with rcrMIP-1 alpha , rhMIP-1 alpha , rmMIP-1 alpha , rcrMIP-1 beta , rmMIP-1 beta , rhMIP-1 beta rhPARC, rcrRANTES, rhRANTES, rmRANTES, rhTARC, or rmTARC, | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction