missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG2a Isotype Control, APC-Alexa Fluor™ 750
Mouse Isotype Control
Brand: Invitrogen™ MG2A27
This item is not returnable.
View return policy
Description
Mouse IgG2a Isotype Control for Flow
Mouse IgG2a Control antibody specifically detects Mouse IgG2a in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG2a | |
APC-Alexa Fluor 750 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG2a | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests | |
Alexa Fluor™ |